Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021120.3 | NP_066943.2 | 7 - 8 | 1490 | 72 | |
XM_006724625.2 | XP_006724688.1 | 7 - 8 | 1495 | 72 | ||
XM_006724626.2 | XP_006724689.1 | 7 - 8 | 1495 | 72 | ||
XM_011530883.1 | XP_011529185.1 | 7 - 8 | 1495 | 72 | ||
GenBank mRNA | AB209328.1 | - | 7 - 8 | 1490 | 72 | |
AK315041.1 | - | 7 - 8 | 1429 | 72 | ||
BC093864.1 | - | 7 - 8 | 1191 | 7939709 | 72 | |
BC093866.1 | - | 7 - 8 | 1191 | 7939711 | 72 | |
U49089.1 | - | 7 - 8 | 1376 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |