Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000323327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000323327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000323327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000323327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000323327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000323327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
NM_153714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
NM_153714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
NM_153714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
NM_153714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
NM_153714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
NM_153714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010039.hg.1 |
NM_153714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000323327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000323327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000323327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000323327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000323327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000323327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
ENST00000376500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
NM_153714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
NM_153714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
NM_153714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
NM_153714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
NM_153714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
NM_153714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010039.hg.1 |
NM_153714.2 |
Refseq |