Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002967.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002967.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002967.hg.1 |
TC20000235.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002969.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002969.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002969.hg.1 |
TC20000235.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002968.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002968.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002968.hg.1 |
TC20000235.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007236.hg.1 |
ENST00000556017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149184.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149184.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149184.hg.1 |
TC2000007169.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149184.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149184.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149185.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149185.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149185.hg.1 |
TC2000007169.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149185.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149185.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007169.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149186.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149186.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149186.hg.1 |
TC2000007169.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149186.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149186.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007236.hg.1 |
ENST00000556017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149183.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149183.hg.1 |
NM_176812.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149183.hg.1 |
TC2000007169.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149183.hg.1 |
ENST00000217402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149183.hg.1 |
NM_176812.4 |
Refseq |