Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008816.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008816.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008816.hg.1 |
TC20000610.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008344.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158806.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158806.hg.1 |
NM_017545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158806.hg.1 |
TC2000008344.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158806.hg.1 |
ENST00000378789.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158806.hg.1 |
NM_017545.2 |
Refseq |