Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024549.hg.1 |
TC07003364.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024549.hg.1 |
ENST00000265627.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024549.hg.1 |
ENST00000427422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07024549.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000460248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000460248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000460248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000460248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000460248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000475439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000475439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000475439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000475439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000492800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000492800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000492800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000492800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000492800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000492800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
ENST00000492800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700184572.hg.1 |
TC0700011834.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000265627.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000418617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000427422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000442770.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000451904.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000456855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000460248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000460248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000460248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000460248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000460248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000475439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000475439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000475439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000475439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000482624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000492800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000492800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000492800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000492800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000492800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000492800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
ENST00000492800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011834.hg.1 |
NM_000940.2 |
Refseq |