Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007686.hg.1 |
TC05000551.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007686.hg.1 |
ENST00000274458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007686.hg.1 |
ENST00000503424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007686.hg.1 |
ENST00000515539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007686.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007684.hg.1 |
TC05000551.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007684.hg.1 |
ENST00000274458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007684.hg.1 |
ENST00000515539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05007684.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013217.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
ENST00000274458.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
ENST00000506589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
ENST00000507356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
ENST00000515539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
ENST00000632434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
NM_001308080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
NM_016144.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193876.hg.1 |
TC0500013217.hg.1 |
Clariom D TC |