Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013355.hg.1 |
TC01000841.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013355.hg.1 |
ENST00000337338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013355.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013362.hg.1 |
TC01000841.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013362.hg.1 |
ENST00000337338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013362.hg.1 |
ENST00000394593.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013362.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013362.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013363.hg.1 |
TC01000841.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013363.hg.1 |
ENST00000337338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013363.hg.1 |
ENST00000394593.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013363.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013363.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000414841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000414841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000414841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000414841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000525774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000525774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000525774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000525774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000532201.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000532201.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000532201.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
ENST00000532201.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
ENST00000414841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
ENST00000532201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
ENST00000414841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
ENST00000532201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238358.hg.1 |
TC0100018247.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100238360.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238360.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238360.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238360.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238360.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238360.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238360.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238360.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238360.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238360.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238360.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238360.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238360.hg.1 |
TC0100018247.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100238347.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238347.hg.1 |
ENST00000525774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238347.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238347.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238347.hg.1 |
ENST00000525774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238347.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238347.hg.1 |
TC0100018247.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
ENST00000414841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
ENST00000525774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
ENST00000532201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
ENST00000414841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
ENST00000525774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
ENST00000532201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100238357.hg.1 |
TC0100018247.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000337338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000394593.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000414841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000414841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000414841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000414841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000441269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000525774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000525774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000525774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000525774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000527156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000532201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000532201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000532201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
ENST00000532201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_001134479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018247.hg.1 |
NM_018103.4 |
Refseq |