Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000503813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000503813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000512167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000512167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000512167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000512167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000512167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
ENST00000512167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005140.hg.1 |
TC04000378.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005140.hg.1 |
ENST00000451783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005140.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000503813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000503813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000512167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000512167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000512167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000512167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000512167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
ENST00000512167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007773.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153000.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153000.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153000.hg.1 |
ENST00000512167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153000.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153000.hg.1 |
ENST00000451783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153000.hg.1 |
ENST00000503711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153000.hg.1 |
ENST00000512167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153000.hg.1 |
NR_003720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153000.hg.1 |
TC0400007773.hg.1 |
Clariom D TC |