Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008350.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008350.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008350.hg.1 |
TC22000464.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008361.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008361.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008361.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008361.hg.1 |
TC22000464.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000510265.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
ENST00000510265.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008360.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008360.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008360.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008360.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008360.hg.1 |
TC22000464.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008354.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008354.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008354.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008354.hg.1 |
TC22000464.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008353.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008353.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008353.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008353.hg.1 |
TC22000464.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008356.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008356.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008356.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008356.hg.1 |
TC22000464.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008352.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008352.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008352.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008352.hg.1 |
TC22000464.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200169604.hg.1 |
TC2200009307.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200169604.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169604.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169604.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169604.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169604.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169604.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169604.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169604.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169609.hg.1 |
TC2200009307.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200169609.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169609.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169609.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169609.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169609.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169609.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169609.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169609.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169609.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169609.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169615.hg.1 |
TC2200009307.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200169615.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169615.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169615.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169615.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169615.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169615.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169615.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169615.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169615.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169615.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169601.hg.1 |
TC2200009307.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200169601.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169601.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169601.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169601.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169601.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169601.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169607.hg.1 |
TC2200009307.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200169607.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169607.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169607.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169607.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169607.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169607.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169607.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169607.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169607.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169607.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169603.hg.1 |
TC2200009307.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200169603.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169603.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169603.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169603.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169603.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169603.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169603.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169603.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169614.hg.1 |
TC2200009307.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200169614.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169614.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169614.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169614.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169614.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169614.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169614.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169614.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169614.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169614.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169614.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169614.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000338526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000434304.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000456726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000457060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000510265.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
ENST00000510265.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009307.hg.1 |
NR_040115.1 |
Refseq |