Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011766.hg.1 |
ENST00000215912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011766.hg.1 |
ENST00000416925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011766.hg.1 |
ENST00000441972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011766.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011766.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011766.hg.1 |
TC22000667.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011769.hg.1 |
ENST00000215912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011769.hg.1 |
ENST00000416925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011769.hg.1 |
ENST00000441972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011769.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011769.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011769.hg.1 |
TC22000667.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011767.hg.1 |
ENST00000215912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011767.hg.1 |
ENST00000416925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011767.hg.1 |
ENST00000441972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011767.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011767.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011767.hg.1 |
TC22000667.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200160957.hg.1 |
TC2200008475.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160957.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160957.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160957.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160957.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160957.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160957.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160957.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160957.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160957.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160957.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160959.hg.1 |
TC2200008475.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160959.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160959.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160959.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160959.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160959.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160959.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160959.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160959.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160959.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160959.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160959.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160959.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160958.hg.1 |
TC2200008475.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160958.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160958.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160958.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160958.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160958.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160958.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160958.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160958.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160958.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160958.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160958.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160958.hg.1 |
NM_052880.4 |
Refseq |