Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002998.hg.1 |
TC6_cox_hap2000164.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002998.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002908.hg.1 |
TC6_dbb_hap3000153.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002908.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002665.hg.1 |
TC6_mcf_hap5000141.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002665.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002668.hg.1 |
TC6_mcf_hap5000141.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002668.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002665.hg.1 |
TC6_ssto_hap7000136.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002665.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002977.hg.1 |
TC6_qbl_hap6000156.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002977.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017675.hg.1 |
TC06001535.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017675.hg.1 |
ENST00000375858.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017675.hg.1 |
ENST00000375860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017675.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017675.hg.1 |
ENST00000472051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017675.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017679.hg.1 |
TC06001535.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017679.hg.1 |
ENST00000375858.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017679.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017679.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002668.hg.1 |
TC6_ssto_hap7000136.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002668.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000460141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000460141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000460141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000460141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000460141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000461572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000461572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000467098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000467098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000467098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000467098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000467098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000467098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000474678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000474678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000474678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
ENST00000474678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011445.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017678.hg.1 |
TC06001535.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017678.hg.1 |
ENST00000375858.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017678.hg.1 |
ENST00000375860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017678.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017678.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002905.hg.1 |
TC6_dbb_hap3000153.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002905.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003001.hg.1 |
TC6_cox_hap2000164.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003001.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002523.hg.1 |
TC6_mann_hap4000141.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002523.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002980.hg.1 |
TC6_qbl_hap6000156.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002980.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002526.hg.1 |
TC6_mann_hap4000141.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002526.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000460141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000460141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000460141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000460141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000460141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000461572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000461572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000467098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000467098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000467098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000467098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000467098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000467098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000474678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000474678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000474678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
ENST00000474678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011445.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
ENST00000460141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
ENST00000467098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
ENST00000474678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
ENST00000375863.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
ENST00000460141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
ENST00000467098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
ENST00000474395.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
ENST00000474678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177440.hg.1 |
TC0600011445.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600177443.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177443.hg.1 |
ENST00000460141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177443.hg.1 |
ENST00000474678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177443.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177443.hg.1 |
ENST00000383237.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177443.hg.1 |
ENST00000460141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177443.hg.1 |
ENST00000474678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177443.hg.1 |
NM_025262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177443.hg.1 |
TC0600011445.hg.1 |
Clariom D TC |