Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012393.hg.1 |
ENST00000283025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012393.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012393.hg.1 |
TC16000858.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000574923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000574923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000574923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000574923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000574923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000576638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000576638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000576638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000576638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000576638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
ENST00000576638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600171756.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171756.hg.1 |
ENST00000576638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171756.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600171756.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171756.hg.1 |
ENST00000576638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171756.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600171756.hg.1 |
TC1600009367.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000283025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000574923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000574923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000574923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000574923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000574923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000576638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000576638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000576638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000576638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000576638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
ENST00000576638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009367.hg.1 |
NM_144674.1 |
Refseq |