Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13010067.hg.1 |
ENST00000335288.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13010067.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13010067.hg.1 |
TC13000897.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300166041.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166041.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300166041.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166041.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300166041.hg.1 |
TC1300009919.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |