Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000298173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000298173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000298173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000298173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000298173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000298173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000298173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000298173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000298173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000298173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000298173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000298173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000298173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000298173.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000434192.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000434192.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000434192.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000434192.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000434192.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000434192.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000434192.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000434192.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000434192.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000553612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000556268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000556268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000556268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000556268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000556268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000556268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
ENST00000556268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_001278940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_015859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_201595.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_201595.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_201595.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_201595.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_201595.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_201595.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_201595.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_201595.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_201595.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_201595.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_201595.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_201595.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009834.hg.1 |
NM_201595.2 |
Refseq |