Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026503.hg.1 |
TC06002112.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026503.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026503.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013186.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191040.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191040.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191040.hg.1 |
ENST00000367976.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191040.hg.1 |
NM_001901.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191040.hg.1 |
TC0600013186.hg.1 |
Clariom D TC |