Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000011684.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000304581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000304581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000304581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000304581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000304581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000304581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000304581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000396988.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000449001.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000535616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000535616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000535616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000536531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000538208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000538208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000538208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000538208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000543000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000543000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000543000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000544743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000544743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
ENST00000544743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_001144857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006610.hg.1 |
NM_018173.3 |
Refseq |