Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_173853_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8040957 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8040955 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8040959 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8040951 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8040950 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8040956 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8040953 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8040954 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8040958 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8040949 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
235148_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11729482_a_at |
- |
- |
Affymetrix Human Genome U219 Array |
11729482_PM_a_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000452499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000452499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000452499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000494572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000494572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000494572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000494572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000494572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000494572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000494572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002359.hg.1 |
TC02000175.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002359.hg.1 |
ENST00000543753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002359.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148532.hg.1 |
ENST00000452499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148532.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148532.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148532.hg.1 |
ENST00000452499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148532.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148532.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148532.hg.1 |
TC0200007068.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000288873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000407293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000452499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000452499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000452499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000453171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000464699.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000494572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000494572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000494572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000494572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000494572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000494572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000494572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
ENST00000543753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_001168364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007068.hg.1 |
NM_173853.3 |
Refseq |
Human Genome U133 Plus 2.0 Array |
235148_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
235148_at |
- |
- |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_173853_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
235148_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
235148_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
235148_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
235148_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
235148_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
235148_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
235148_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
235148_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
235148_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
235148_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
235148_at4 |
- |
- |
Human Genome U95 Set |
51879_at |
- |
- |
Human Genome U95 Set |
51879_PM_at |
- |
- |
Human Genome U133 Set |
235148_at |
- |
- |
Human Genome U133 Set |
235148_PM_at |
- |
- |