Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035880.hg.1 |
TC02002211.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035880.hg.1 |
ENST00000341068.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035880.hg.1 |
NM_022662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201862.hg.1 |
ENST00000341068.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201862.hg.1 |
ENST00000427997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201862.hg.1 |
NM_022662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201862.hg.1 |
ENST00000341068.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201862.hg.1 |
ENST00000427997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201862.hg.1 |
NM_022662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201862.hg.1 |
TC0200013894.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008302.hg.1 |
ENST00000454465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008319.hg.1 |
ENST00000427434.1 |
Ensembl |