Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297419.1 | NP_001284348.1 | 2 - 3 | 342 | 60 | |
NM_001297420.1 | NP_001284349.1 | 1 - 2 | 109 | 60 | ||
NM_001297422.1 | NP_001284351.1 | 2 - 3 | 230 | 60 | ||
NM_134424.3 | NP_602296.2 | 2 - 3 | 230 | 60 | ||
NR_123713.1 | 3 - 4 | 306 | 60 | |||
XM_005253720.4 | XP_005253777.1 | 3 - 4 | 1104 | 60 | ||
XM_005253721.2 | XP_005253778.1 | 2 - 3 | 330 | 60 | ||
XM_011520990.2 | XP_011519292.1 | 3 - 4 | 366 | 60 | ||
XM_011520991.2 | XP_011519293.1 | 2 - 3 | 232 | 60 | ||
XM_011520993.2 | XP_011519295.1 | 2 - 3 | 181 | 60 | ||
XM_017019769.1 | XP_016875258.1 | 3 - 4 | 487 | 60 | ||
XM_017019770.1 | XP_016875259.1 | 2 - 3 | 181 | 60 | ||
XM_017019771.1 | XP_016875260.1 | 2 - 3 | 181 | 60 | ||
GenBank mRNA | AF125948.1 | - | 2 - 3 | 134 | 60 | |
AF125949.1 | - | 2 - 3 | 202 | 60 | ||
AF125950.1 | - | 1 - 2 | 91 | 60 | ||
AK290047.1 | - | 2 - 3 | 150 | 60 | ||
AK292160.1 | - | 2 - 3 | 174 | 60 | ||
AK294709.1 | - | 2 - 3 | 175 | 60 | ||
AK312026.1 | - | 2 - 3 | 138 | 60 | ||
AY540753.1 | - | 1 - 2 | 92 | 60 | ||
BC104015.1 | - | 2 - 3 | 173 | 60 | ||
BC104016.1 | - | 2 - 3 | 173 | 60 | ||
BC104017.1 | - | 2 - 3 | 173 | 60 | ||
BC114507.1 | - | 2 - 3 | 173 | 60 | ||
BC114954.1 | - | 2 - 3 | 169 | 60 | ||
BC144458.1 | - | 1 - 2 | 115 | 60 | ||
HY045122.1 | - | 3 - 4 | 369 | 60 | ||
L33262.1 | - | 2 - 3 | 360 | 60 | ||
U12134.1 | - | 1 - 2 | 123 | 60 | ||
U27516.1 | - | 1 - 2 | 122 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |