Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000089.3 | NP_000080.2 | 13 - 14 | 1112 | 73 | |
GenBank mRNA | AK226074.1 | - | 13 - 14 | 763 | 73 | |
BC042586.1 | - | 13 - 14 | 759 | 4803351 | 73 | |
BC054498.1 | - | 13 - 14 | 741 | 6474549 | 73 | |
J03464.1 | - | 13 - 14 | 73 | |||
KU177988.1 | - | 13 - 14 | 641 | 73 | ||
KU177989.1 | - | 13 - 14 | 641 | 73 | ||
Y00724.1 | - | 13 - 14 | 717 | 73 | ||
Z74616.1 | - | 13 - 14 | 780 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |