Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000416571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000416571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000416571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000416571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000416571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000416571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000416571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000422266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000422266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000422266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000422266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000422266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000425072.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000425072.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000425072.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000425072.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000425072.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000492462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000492462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000492462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000492462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000492462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000494654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000494654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000494654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000613547.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000613547.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000613547.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000613547.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000613547.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000613547.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011379.hg.1 |
ENST00000613547.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176734.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176734.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176734.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176734.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176734.hg.1 |
TC0600011379.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600176735.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176735.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176735.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176735.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176735.hg.1 |
TC0600011379.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600176738.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176738.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176738.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176738.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176738.hg.1 |
TC0600011379.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000416571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000416571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000416571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000416571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000416571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000416571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000416571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000417033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000422266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000422266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000422266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000422266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000422266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000425072.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000425072.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000425072.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000425072.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000425072.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000492462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000492462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000492462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000492462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000492462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000494654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000494654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000494654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000613547.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000613547.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000613547.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000613547.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000613547.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000613547.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011379.hg.1 |
ENST00000613547.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176733.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176733.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176733.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176733.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176733.hg.1 |
TC0600011379.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600176736.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176736.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176736.hg.1 |
ENST00000376406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176736.hg.1 |
ENST00000489540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176736.hg.1 |
TC0600011379.hg.1 |
Clariom D TC |