Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018643.hg.1 |
TC05001319.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018643.hg.1 |
ENST00000306862.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018643.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000503655.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000503655.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000503655.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000503655.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000506213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000506213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000506213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000506213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000506213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000506213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000509489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000509489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000509489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000509489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000514462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000514462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000514462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000514462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000514462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
ENST00000514462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174324.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174324.hg.1 |
ENST00000506213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174324.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174324.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174324.hg.1 |
ENST00000506213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174324.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174324.hg.1 |
TC0500010641.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000306862.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000503655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000503655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000503655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000503655.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000506213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000506213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000506213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000506213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000506213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000506213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000509489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000509489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000509489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000509489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000514462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000514462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000514462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000514462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000514462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
ENST00000514462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010641.hg.1 |
NM_198566.3 |
Refseq |