Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KITLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000899.4 | NP_000890.1 | 8 - 9 | 982 | 138 | |
NM_003994.5 | NP_003985.2 | 7 - 8 | 898 | 138 | ||
GenBank mRNA | AF400436.1 | - | 8 - 9 | 970 | 139 | |
AK290319.1 | - | 7 - 8 | 894 | 138 | ||
AK293002.1 | - | 7 - 8 | 814 | 138 | ||
BC069733.1 | - | 8 - 9 | 943 | 7262647 | 138 | |
BC069783.1 | - | 8 - 9 | 944 | 7262659 | 138 | |
BC069797.1 | - | 8 - 9 | 939 | 7262576 | 138 | |
BC074725.2 | - | 8 - 9 | 827 | 30915176 | 138 | |
BC126166.1 | - | 8 - 9 | 937 | 8991882 | 138 | |
BC143899.1 | - | 7 - 8 | 853 | 9052417 | 138 | |
CR749222.1 | - | 7 - 8 | 730 | 138 | ||
M59964.1 | - | 8 - 9 | 970 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |