Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012888.hg.1 |
TC09000982.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012888.hg.1 |
ENST00000333916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012888.hg.1 |
ENST00000495958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012888.hg.1 |
ENST00000535437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012888.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012888.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000444762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000495958.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000495958.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000495958.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000495958.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000495958.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000495958.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000517946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000517946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000517946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000517946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000520187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000520187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000520187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000520187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000520187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000520187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000520187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000444762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000495958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000495958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000495958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000495958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000495958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000495958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000517946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000517946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000517946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000517946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000520187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000520187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000520187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000520187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000520187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000520187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000520187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009768.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
ENST00000495958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
ENST00000517946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
ENST00000333916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
ENST00000483493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
ENST00000495958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
ENST00000517946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
ENST00000625311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
NM_001167575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
NM_024828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166783.hg.1 |
TC0900009768.hg.1 |
Clariom D TC |