Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021029.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021029.hg.1 |
ENST00000370033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021029.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021029.hg.1 |
TC10001615.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000410498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000456149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000456149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000456149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000456149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000470043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000470043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000470043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000470043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000470043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000470043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000495001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000495001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000495001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000495001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
ENST00000495001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179846.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179846.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179846.hg.1 |
ENST00000470043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179846.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179846.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179846.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179846.hg.1 |
ENST00000470043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179846.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179846.hg.1 |
TC1000011662.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000311122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000370033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000410498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000456149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000456149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000456149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000456149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000470043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000470043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000470043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000470043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000470043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000470043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000495001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000495001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000495001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000495001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
ENST00000495001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011662.hg.1 |
NM_024541.2 |
Refseq |