Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009540.hg.1 |
ENST00000604297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009540.hg.1 |
ENST00000604297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368653.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000368654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000464771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000464771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000478293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000478293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000478293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000478293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000484853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000484853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000484853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000484853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
ENST00000617118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_001145966.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012235.hg.1 |
NM_002417.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009540.hg.1 |
ENST00000604297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009540.hg.1 |
ENST00000604297.1 |
Ensembl |