Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024043.hg.1 |
TC06001952.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024043.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024043.hg.1 |
ENST00000369303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024043.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024046.hg.1 |
TC06001952.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024046.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024042.hg.1 |
TC06001952.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024042.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024042.hg.1 |
ENST00000369303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024042.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024044.hg.1 |
TC06001952.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024044.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024044.hg.1 |
ENST00000369303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024044.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186831.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186831.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186831.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186831.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186831.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186831.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186831.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186831.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186831.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186831.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186831.hg.1 |
TC0600012597.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186832.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186832.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186832.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186832.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186832.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186832.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186832.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186832.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186832.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186832.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186832.hg.1 |
TC0600012597.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186833.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186833.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186833.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186833.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186833.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186833.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186833.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186833.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186833.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186833.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186833.hg.1 |
TC0600012597.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012597.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186830.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186830.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186830.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186830.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186830.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186830.hg.1 |
ENST00000369297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186830.hg.1 |
ENST00000369303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186830.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186830.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186830.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186830.hg.1 |
TC0600012597.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186835.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186835.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186835.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186835.hg.1 |
NM_001288629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186835.hg.1 |
NM_001288630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186835.hg.1 |
NM_004440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186835.hg.1 |
TC0600012597.hg.1 |
Clariom D TC |