Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011252.hg.1 |
TC0X000827.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011252.hg.1 |
ENST00000381042.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011252.hg.1 |
ENST00000444736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011252.hg.1 |
ENST00000537427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011252.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011252.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011252.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000442940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000442940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000442940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000442940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000442940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000442940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006649.hg.1 |
ENST00000458328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006649.hg.1 |
ENST00000458328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006649.hg.1 |
ENST00000458328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
ENST00000442940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
ENST00000442940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161851.hg.1 |
TC0X00009018.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000381042.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000442940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000442940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000442940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000442940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000442940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000442940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000444736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
ENST00000537427.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001142389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_001172672.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009018.hg.1 |
NM_004650.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006649.hg.1 |
ENST00000458328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006649.hg.1 |
ENST00000458328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006649.hg.1 |
ENST00000458328.1 |
Ensembl |