Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000379774.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000379774.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000379774.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000379774.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000379774.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000468843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000468843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000468843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000468843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000468843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000468843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000527582.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000527582.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000527582.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000527582.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000609540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000609540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000609540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
ENST00000609540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
NM_020233.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
NM_020233.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
NM_020233.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
NM_020233.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
NM_020233.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006877.hg.1 |
NM_020233.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700148442.hg.1 |
ENST00000379774.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148442.hg.1 |
ENST00000468843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148442.hg.1 |
ENST00000609540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148442.hg.1 |
NM_020233.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700148442.hg.1 |
ENST00000379774.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148442.hg.1 |
ENST00000468843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148442.hg.1 |
ENST00000609540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148442.hg.1 |
NM_020233.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700148442.hg.1 |
TC1700006877.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000379774.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000379774.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000379774.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000379774.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000379774.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000468843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000468843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000468843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000468843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000468843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000468843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000527582.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000527582.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000527582.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000527582.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000609540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000609540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000609540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
ENST00000609540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
NM_020233.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
NM_020233.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
NM_020233.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
NM_020233.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
NM_020233.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006877.hg.1 |
NM_020233.4 |
Refseq |