Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009765.hg.1 |
TC09000760.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009765.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009765.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009764.hg.1 |
TC09000760.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009764.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009764.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000483873.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000483873.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000483873.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000483873.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000483873.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000483873.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
ENST00000483873.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009761.hg.1 |
TC09000760.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009761.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009761.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009763.hg.1 |
TC09000760.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009763.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009763.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000483873.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000483873.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000483873.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000483873.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000483873.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000483873.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
ENST00000483873.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009032.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161450.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161450.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161450.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161450.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161450.hg.1 |
TC0900009032.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900161453.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161453.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161453.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161453.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161453.hg.1 |
TC0900009032.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900161449.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161449.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161449.hg.1 |
ENST00000372146.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161449.hg.1 |
NM_012204.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161449.hg.1 |
TC0900009032.hg.1 |
Clariom D TC |