Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011533394.2 | XP_011531696.1 | 1 - 2 | 765 | 130 | |
XM_011533396.2 | XP_011531698.1 | 1 - 2 | 482 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |