Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002217.3 | NP_002208.3 | 16 - 17 | 1942 | 74 | |
XM_005265105.4 | XP_005265162.1 | 17 - 18 | 2053 | 74 | ||
XM_017006356.1 | XP_016861845.1 | 15 - 16 | 1848 | 74 | ||
GenBank mRNA | AK222757.1 | - | 16 - 17 | 1914 | 74 | |
AK223483.1 | - | 16 - 17 | 1940 | 74 | ||
BC107604.1 | - | 16 - 17 | 1912 | 40035241 | 74 | |
BC107605.1 | - | 16 - 17 | 1912 | 40035249 | 74 | |
BC107814.1 | - | 16 - 17 | 1912 | 40035238 | 74 | |
X67055.1 | - | 16 - 17 | 1914 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |