Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003230.hg.1 |
ENST00000347644.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003230.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003230.hg.1 |
TC15000354.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000560438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000560438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000560438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000560438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000560438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000560438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000560767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000560767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000560767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000560767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000560767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000560767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
ENST00000560767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003229.hg.1 |
ENST00000347644.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003229.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003229.hg.1 |
TC15000354.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000559924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000560438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000560438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000560438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000560438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000560438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000560438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000560767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000560767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000560767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000560767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000560767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000560767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
ENST00000560767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007116.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150630.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150630.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150630.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150630.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150630.hg.1 |
TC1500007116.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500150629.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150629.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150629.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150629.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150629.hg.1 |
TC1500007116.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500150628.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150628.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150628.hg.1 |
ENST00000347644.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150628.hg.1 |
NM_004212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150628.hg.1 |
TC1500007116.hg.1 |
Clariom D TC |