Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015102.hg.1 |
TC12003270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015102.hg.1 |
ENST00000229243.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015102.hg.1 |
ENST00000414226.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015102.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015102.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000535884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000535884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000535884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000535884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000535884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000535884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000535884.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000538524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000538524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000538524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000538524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000540513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000540513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000540513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000540513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000540513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000544352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000544352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000544352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000544352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000544352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000544352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000546114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000546114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000546114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000546114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000546114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
ENST00000546114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172951.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172951.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172951.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172951.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172951.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172951.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172951.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172951.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172951.hg.1 |
TC1200009746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000229243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000414226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000535884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000535884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000535884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000535884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000535884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000535884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000535884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000536350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000538524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000538524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000538524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000538524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000540513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000540513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000540513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000540513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000540513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000542357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000544352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000544352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000544352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000544352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000544352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000544352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000546114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000546114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000546114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000546114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000546114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
ENST00000546114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009746.hg.1 |
NM_032489.2 |
Refseq |