Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004184.hg.1 |
ENST00000219919.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004184.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004184.hg.1 |
ENST00000558772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004184.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004184.hg.1 |
TC15000441.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000559443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000559443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000559443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
ENST00000559443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500152465.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152465.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152465.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152465.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500152465.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152465.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152465.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500152465.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500152465.hg.1 |
TC1500007355.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000219919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000536493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000558772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000559443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000559443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000559443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
ENST00000559443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007355.hg.1 |
NM_020980.4 |
Refseq |