Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243835.1 | NP_001230764.1 | 11 - 12 | 1352 | 147 | |
NM_003151.3 | NP_003142.1 | 11 - 12 | 1403 | 147 | ||
XM_006712719.3 | XP_006712782.1 | 11 - 12 | 1398 | 147 | ||
XM_011511705.2 | XP_011510007.1 | 14 - 15 | 1549 | 147 | ||
XM_017004784.1 | XP_016860273.1 | 11 - 12 | 1157 | 147 | ||
GenBank mRNA | AK300868.1 | - | 11 - 12 | 1156 | 147 | |
AK301172.1 | - | 9 - 10 | 827 | 147 | ||
BC031212.1 | - | 11 - 12 | 1324 | 4830583 | 147 | |
L78440.1 | - | 11 - 12 | 1169 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |