Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011001.hg.1 |
TC09000837.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011001.hg.1 |
ENST00000409858.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011001.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011000.hg.1 |
TC09000837.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011000.hg.1 |
ENST00000409858.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011000.hg.1 |
ENST00000476184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011000.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011002.hg.1 |
TC09000837.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011002.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011002.hg.1 |
ENST00000409858.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011002.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000476184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000476184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000476184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
ENST00000476184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010999.hg.1 |
TC09000837.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010999.hg.1 |
ENST00000409858.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010999.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163344.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163344.hg.1 |
ENST00000476184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163344.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163344.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163344.hg.1 |
ENST00000476184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163344.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163344.hg.1 |
TC0900009259.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900163342.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163342.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163342.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163342.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163342.hg.1 |
TC0900009259.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900163345.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163345.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163345.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163345.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163345.hg.1 |
TC0900009259.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000474787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000476184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000476184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000476184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
ENST00000476184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009259.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163346.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163346.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163346.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163346.hg.1 |
ENST00000360271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163346.hg.1 |
ENST00000409858.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163346.hg.1 |
NM_207309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163346.hg.1 |
TC0900009259.hg.1 |
Clariom D TC |