Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000541.4 | NP_000532.2 | 14 - 16 | 1492 | 90 | |
XM_011511593.2 | XP_011509895.1 | 10 - 12 | 1196 | 90 | ||
XM_011511594.1 | XP_011509896.1 | 10 - 12 | 805 | 90 | ||
XM_011511596.1 | XP_011509898.1 | 11 - 13 | 1399 | 90 | ||
GenBank mRNA | BQ636894.1 | - | 6 - 8 | 406 | 90 | |
BU734369.1 | - | 1 - 3 | 90 | |||
BX641043.1 | - | 12 - 14 | 2323 | 90 | ||
BX647821.1 | - | 14 - 16 | 3136 | 90 | ||
BX647827.1 | - | 11 - 13 | 6290 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens |