Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000393613.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000393613.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000393613.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000393613.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000393613.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000488792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000488792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000488792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000488792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
ENST00000488792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010740.hg.1 |
TC07000688.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010740.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010740.hg.1 |
ENST00000539794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010740.hg.1 |
ENST00000543645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010740.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000393613.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000393613.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000393613.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000393613.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000393613.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000488792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000488792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000488792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000488792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
ENST00000488792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700161745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161745.hg.1 |
ENST00000265717.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700161745.hg.1 |
NM_002736.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161745.hg.1 |
TC0700008745.hg.1 |
Clariom D TC |