Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000312350.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000312350.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000312350.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000312350.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000312350.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000312350.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000312350.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000312350.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000312350.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000312350.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000312350.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000319231.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000319231.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000319231.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000319231.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000319231.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000319231.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000319231.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000319231.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000319231.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000319231.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000340700.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000340700.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000340700.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000340700.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000340700.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000340700.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000340700.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000340700.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000340700.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000340700.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000340700.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000556282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000556282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
ENST00000556282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_021161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_021161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_021161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_021161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_021161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_021161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_021161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_021161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_021161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_021161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_021161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138317.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009919.hg.1 |
NM_138318.2 |
Refseq |