Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004562.2 | NP_004553.2 | 5 - 6 | 752 | 100 | |
XM_011535863.1 | XP_011534165.1 | 5 - 6 | 749 | 100 | ||
XM_017010908.1 | XP_016866397.1 | 5 - 6 | 1666 | 100 | ||
XM_017010909.1 | XP_016866398.1 | 4 - 5 | 546 | 100 | ||
GenBank mRNA | AK292590.1 | - | 5 - 6 | 713 | 100 | |
BC022014.2 | - | 5 - 6 | 716 | 4824892 | 100 | |
EF375726.1 | - | 4 - 5 | 618 | 100 | ||
GU345837.1 | - | 4 - 5 | 471 | 100 | ||
GU345838.1 | - | 3 - 4 | 513 | 100 | ||
GU345840.1 | - | 4 - 5 | 635 | 100 | ||
GU357501.1 | - | 2 - 3 | 108 | 100 | ||
GU357502.1 | - | 1 - 2 | 108 | 100 | ||
GU361466.1 | - | 3 - 4 | 513 | 100 | ||
GU361468.1 | - | 3 - 4 | 394 | 100 | ||
GU361469.1 | - | 5 - 6 | 732 | 100 | ||
GU361470.1 | - | 5 - 6 | 733 | 100 | ||
KC357595.1 | - | 7 - 8 | 830 | 100 | ||
KJ534911.1 | - | 4 - 5 | 618 | 100 | ||
KJ534912.1 | - | 4 - 5 | 618 | 100 | ||
KU178240.1 | - | 4 - 5 | 618 | 100 | ||
KU178241.1 | - | 4 - 5 | 618 | 100 | ||
KU178242.1 | - | 4 - 5 | 615 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |