Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018940.hg.1 |
TC02001235.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018940.hg.1 |
ENST00000264380.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018940.hg.1 |
ENST00000392200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018940.hg.1 |
NM_015040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200175300.hg.1 |
ENST00000264380.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200175300.hg.1 |
NM_015040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200175300.hg.1 |
ENST00000264380.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200175300.hg.1 |
NM_015040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200175300.hg.1 |
TC0200010624.hg.1 |
Clariom D TC |