Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009394.hg.1 |
ENST00000369701.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009394.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009394.hg.1 |
TC10000788.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000393176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000393176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000393176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000393176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
ENST00000393176.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160321.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160321.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160321.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160321.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160321.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160321.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160321.hg.1 |
TC1000008813.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369699.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000369701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000393176.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000393176.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000393176.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000393176.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
ENST00000393176.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008813.hg.1 |
NM_014978.2 |
Refseq |