Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002617.hg.1 |
TC02000184.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002617.hg.1 |
ENST00000327757.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002617.hg.1 |
ENST00000422425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002617.hg.1 |
NM_001170585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002617.hg.1 |
NM_153021.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148888.hg.1 |
ENST00000327757.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148888.hg.1 |
ENST00000404858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148888.hg.1 |
ENST00000422425.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148888.hg.1 |
NM_001170585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148888.hg.1 |
NM_153021.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148888.hg.1 |
ENST00000327757.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148888.hg.1 |
ENST00000404858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148888.hg.1 |
ENST00000422425.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148888.hg.1 |
NM_001170585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148888.hg.1 |
NM_153021.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148888.hg.1 |
TC0200007102.hg.1 |
Clariom D TC |