Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005923.3 | NP_005914.1 | 12 - 13 | 2187 | 112 | |
XM_011535839.2 | XP_011534141.2 | 13 - 14 | 2511 | 112 | ||
XM_017010870.1 | XP_016866359.1 | 13 - 14 | 2510 | 112 | ||
XM_017010871.1 | XP_016866360.1 | 13 - 14 | 2510 | 112 | ||
XM_017010872.1 | XP_016866361.1 | 13 - 14 | 2509 | 112 | ||
XM_017010873.1 | XP_016866362.1 | 13 - 14 | 2508 | 112 | ||
XM_017010876.1 | XP_016866365.1 | 11 - 12 | 1518 | 112 | ||
XM_017010877.1 | XP_016866366.1 | 11 - 12 | 1755 | 112 | ||
GenBank mRNA | AB209093.1 | - | 12 - 13 | 2291 | 112 | |
AK293953.1 | - | 12 - 13 | 1447 | 112 | ||
AK294507.1 | - | 3 - 4 | 303 | 112 | ||
BC054503.2 | - | 12 - 13 | 2134 | 6143181 | 112 | |
BC088829.1 | - | 12 - 13 | 2188 | 6007002 | 112 | |
D84476.2 | - | 12 - 13 | 2093 | 112 | ||
U67156.1 | - | 12 - 13 | 2186 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Homo sapiens Probe spans exons |