Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027050.hg.1 |
TC06002146.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027050.hg.1 |
ENST00000355845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027050.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027050.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007083.hg.1 |
ENST00000518031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007083.hg.1 |
ENST00000518031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007083.hg.1 |
ENST00000518031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007083.hg.1 |
ENST00000518031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006561.hg.1 |
ENST00000623728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191932.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191932.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191932.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191932.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191932.hg.1 |
TC0600013280.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800007083.hg.1 |
ENST00000518031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007083.hg.1 |
ENST00000518031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007083.hg.1 |
ENST00000518031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007083.hg.1 |
ENST00000518031.1 |
Ensembl |