Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000419619.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000419619.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000419619.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000439288.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000439288.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000439288.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000465319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000465319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000465319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000465319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000612036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000612036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000612036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000612036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000612036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000612036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
ENST00000612036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010499.hg.1 |
TC06000889.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010499.hg.1 |
ENST00000368850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010499.hg.1 |
ENST00000368851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010499.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000419619.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000419619.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000419619.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000439288.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000439288.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000439288.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000465319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000465319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000465319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000465319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000612036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000612036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000612036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000612036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000612036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000612036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
ENST00000612036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009143.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162788.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162788.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162788.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162788.hg.1 |
ENST00000368850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162788.hg.1 |
ENST00000368851.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162788.hg.1 |
NM_018593.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162788.hg.1 |
TC0600009143.hg.1 |
Clariom D TC |