Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023040.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023040.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023040.hg.1 |
TC10001738.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012161.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183206.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183206.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183206.hg.1 |
ENST00000368808.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183206.hg.1 |
NM_147191.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183206.hg.1 |
TC1000012161.hg.1 |
Clariom D TC |