Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020081.hg.1 |
TC08001528.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020081.hg.1 |
ENST00000239690.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020081.hg.1 |
ENST00000427660.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020081.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020081.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020080.hg.1 |
TC08001528.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020080.hg.1 |
ENST00000239690.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020080.hg.1 |
ENST00000427660.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020080.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020080.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178056.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178056.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178056.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178056.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178056.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178056.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178056.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178056.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178056.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178056.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178056.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178056.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178056.hg.1 |
TC0800011472.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011472.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178057.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178057.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178057.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178057.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178057.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178057.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178057.hg.1 |
ENST00000239690.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178057.hg.1 |
ENST00000427660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178057.hg.1 |
ENST00000519607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178057.hg.1 |
ENST00000521439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178057.hg.1 |
NM_001128211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178057.hg.1 |
NM_032869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178057.hg.1 |
TC0800011472.hg.1 |
Clariom D TC |