Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007578.hg.1 |
TC06000624.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007578.hg.1 |
ENST00000321037.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007578.hg.1 |
ENST00000371401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007578.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007582.hg.1 |
TC06000624.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007582.hg.1 |
ENST00000321037.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007582.hg.1 |
ENST00000371401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007582.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007577.hg.1 |
TC06000624.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007577.hg.1 |
ENST00000321037.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007577.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157090.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157090.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157090.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157090.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157090.hg.1 |
TC0600008156.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600157089.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157089.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157089.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157089.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157089.hg.1 |
TC0600008156.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600157088.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157088.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157088.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157088.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157088.hg.1 |
TC0600008156.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
ENST00000321037.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008156.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157087.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157087.hg.1 |
NM_014936.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157087.hg.1 |
TC0600008156.hg.1 |
Clariom D TC |